Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, ACS Reagent Grade, ASTM Type I, Ricca Chemical
BIOPHARMASDP

Catalog Number 91501
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
55 gal.
Packaging:
Cubitainer
Hard Pack
Natural Poly Bottle
Poly Drum
7732-18-5
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
962
CHEBI:15377
O
7732-18-5
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
962
CHEBI:15377
O
Color Colorless
Melting Point 0°C
CAS Min % 98.0
Boiling Point 100°C
CAS Max % 100.0
Conductivity max. 0.056
Synonym dihydrogen oxide, dihydrogen monoxide
Solubility Information Miscible
IUPAC Name oxidane
Molecular Weight (g/mol) 18.015
Formula Weight 18.01
Grade ACS
Packaging Natural Poly Bottle
Sterility Non-sterile
Quantity 4 L
Physical Form Liquid
Chemical Name or Material Water, ASTM Type I
Show More Show Less
ShelfLife 18 months
DOTInformation Not Regulated by DOT
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit