Water, ACS Reagent Grade, ASTM Type I, Ricca Chemical

Manufacturer: Ricca Chemical CompanySupplier Diversity Partner 915055

Packaging Drum
Quantity 55 gal.

 View more versions of this product

Catalog No. 13-622-621

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7732-18-5
    CAS Min % 98.0
    CAS Max % 100.0
    Formula Weight 18.01
    Physical State Liquid
    Color Colorless
    Solubility Miscible
    Melting Point 0°C
    Boiling Point 100°C
    Chemical Name or Material Water, ASTM Type I
    Grade ACS Reagent Grade

    Specifications

    Formula Weight 18.01
    Quantity 55 gal.
    Physical State Liquid
    Color Colorless
    Boiling Point 100°C
    Shelf Life 18 months
    Solubility Miscible
    DOT Information Not Regulated by DOT
    Melting Point 0°C
    CAS 7732-18-5
    CAS Min % 98.0
    CAS Max % 100.0
    Packaging Drum
    Chemical Name or Material Water, ASTM Type I
    Grade ACS Reagent Grade

    18 months

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit