Learn More
Andwin Scientific Water ASTM Type II, 5 gal
SDP
Supplier:  Andwin Scientific BDH11685GL
Encompass_Preferred
Deionized, Used for general laboratory applications, and Suitable for feeding ultra-pure systems, among other uses. Ion exchange, distillation, or reverse osmosis and organic adsorption may be required prior to distillation if the purity cannot be attained by single distillation.
Catalog No. 50-112-6067