Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, ASTM Type II, Reagents

Catalog No. 01336890
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
200 L

Catalog No. 01-336-890

Supplier: Reagents Holdings Llc C2346200500A

Only null left
Add to Cart
Add to Cart

Water, ASTM Type II. ACS Reagent Meets ASTM requirements for Distilled Water, ASTM, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Dihydrogen Oxide
IUPAC Name water
SMILES O

Specifications

Color Colorless
Melting Point 0°C
Boiling Point 100°C
Percent Purity 100%
Packaging Poly Bottle
Sterility Non-sterile
Quantity 500 mL
Specific Gravity 1
Physical Form Liquid
Chemical Name or Material Water, ASTM Type II / ACS Reagent Meets ASTM requirements for Distilled Water
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.