Learn More
Chemglass Life Sciences 15L Water Bath, Shallow, General Purpose, Digital, 120V
SDP
Supplier:  Chemglass Life Sciences CLS4950014
Encompass_Preferred
15L Water Bath, Shallow, General Purpose, Digital, 120V. The CLS-4950 Series high-performance water baths are accurate, easy to use, safe and durable. The digital set / digital read P.I.D. temperature controller maximizes rapid heat-up without temperature overshoot. A microprocessor achieves precise temperature control regardless of how the unit is loaded. It has a unique lid design that allows condensation to drain back into the tank without spilling. There is a gasket around the water bath tank to prevent water from dripping onto the controls. A warm air jacketed design radiates heat to the sides & bottom of the tank, eliminating hot spots. The unit has a non-contact, recessed heating element that will prevent burnouts. The water bath tanks are constructed of stainless steel and do not have seams that could leak. The unit's housings have a high quality textured powder coat finish that eliminates rust & chipping and is resistant to nicks, scratches & corrosives.
Catalog No. 50-143-577
Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.