Learn More
Quality Biological IncSupplier Diversity Partner Water, Cell Culture Grade, 4 Liters DFS Item
Manufacturer:  Quality Biological IncSupplier Diversity Partner 118162491
QBI’s Cell Culture Grade Water is high quality water for use as a solvent in the preparation of cell culture media and laboratory reagents. • Endotoxin < 0.03 EU/mL • Mycoplasma Tested • Also used to dilute buffers and suspend cells. • Prepared by reverse osmosis • Passed through fine carbon • Deionized through two resin beds • Serially filtered twice through 0.1 µm positively charged membranes • Endotoxin level < 0.03 EU/mL. • Storage Condition: 15°C to 30°C • Shipping Condition: ambient temperature • Sterile Filtered • Endotoxin and Mycoplasma Tested • Product and/or packaging contains recycled/renewable content • Product and/or packaging is recyclable or reduces waste • Product and/or packaging is safer to human and environmental health • UNSPSC: 41116113 • Harmonization Code: 3822.00.5090 • ECCN: EAR99 • Max Shelf Life: 24 months
Catalog No. 50-146-766