Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

American Educational Products Water Cycle Model Activity Set Products for Science Education

Replicate the individual processes of the water cycle — evaporation, condensation and precipitation.

Specifications

For Use With (Application) Help teachers broaden students' understanding of the water cycle and enhance awareness of the various states of water
Grade 4 to 12
View More Specs

Alert:

CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.


Includes: Set of ten 8.5 x 11 in. (21.6 x 27.9cm) full-color study prints and activities guide; heat lamp not included

Products 2
Catalog Number Mfr. No. Includes Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Includes Product Type Price Quantity & Availability  
S90725A Products for Science Education
View Documents
American Educational Products
8882
Set of ten 8.5 x 11 in. full-color study prints and activities guide Water cycle model activity set This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
S90725B Products for Science Education
View Documents
American Educational Products
8884
Model, support, cloud cover and 4 page lesson Water cycle model only This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Part Not Found
N/A
Description

Description

  • Broaden students' understanding of the water cycle
  • Enhance awareness of the various states of water

Specifications

Specifications

Help teachers broaden students' understanding of the water cycle and enhance awareness of the various states of water
4 to 12
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit