Alfa Aesar™ Water, deuterium depleted, <5ppm Deuterium

Manufacturer: Alfa Aesar™ 3647414

Quantity 25g

 View more versions of this product

Catalog No. AA3647414

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 7732-18-5
    Formula Weight 18.02
    Chemical Name or Material Water, Deuterium depleted

    Specifications

    Formula Weight 18.02
    Melting Point 0°C
    Density 1.00g/mL
    Boiling Point 100°C
    Quantity 25g
    Concentration <5ppm Deuterium
    TSCA TSCA
    Physical Form Liquid
    Odor Odorless
    CAS 7732-18-5
    Chemical Name or Material Water, Deuterium depleted

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit