Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Water, deuterium depleted, deuterium 2-3ppm

For NMR use

Manufacturer:  Alfa Aesar™ 4236922

Quantity 100g

 View more versions of this product

Catalog No. AA4236922


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7732-18-5
MFCD00011332
Water, Deuterium depleted, deuterium 2 to 3ppm
18.005
14,10039

Specifications

7732-18-5
18.005
14,10039
100°C
Water, Deuterium depleted, deuterium 2 to 3ppm
Liquid
MFCD00011332
Odorless
100g

Safety and Handling

SDS & Safety Documents

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit