Alfa Aesar™ Water, deuterium depleted, deuterium 2-3ppm

For NMR use

Manufacturer: Alfa Aesar™ 4236914

Quantity 25g

 View more versions of this product

Catalog No. AA4236914

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 7732-18-5
    Formula Weight 18.02
    MDL Number MFCD00011332
    Merck Index 14,10039
    Chemical Name or Material Water, Deuterium depleted, deuterium 2 to 3ppm

    Specifications

    Formula Weight 18.02
    Physical Form Liquid
    MDL Number MFCD00011332
    Boiling Point 100°C
    TSCA TSCA
    CAS 7732-18-5
    Merck Index 14,10039
    Odor Odorless
    Quantity 25g
    Chemical Name or Material Water, Deuterium depleted, deuterium 2 to 3ppm

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit