Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, Extra Pure, Deionized, ACROS Organics™

Click to view available options
Quantity:
5L
25L
Packaging:
Plastic drum
7732-18-5
H2O
18.015
XLYOFNOQVPJJNP-UHFFFAOYSA-N
dihydrogen oxide,dihydrogen monoxide
962
CHEBI:15377
oxidane
O
7732-18-5
H2O
18.015
XLYOFNOQVPJJNP-UHFFFAOYSA-N
dihydrogen oxide,dihydrogen monoxide
962
CHEBI:15377
oxidane
O
Biological buffer
Conductivity 5.0μS/cm Max.
Linear Formula H2O
Fieser 16,383
Merck Index 15, 10237
Formula Weight 18.02
Physical Form Liquid
Grade Extra Pure
Oxidizable Substances Passes test
Packaging Plastic drum
Residue after Evaporation 25ppm max.
Viscosity 0.89 mPa.s (25°C)
Density 1.0000g/mL
Color Colorless
Boiling Point 100.0°C
Quantity 5L
Refractive Index 1.3334
Specific Gravity 1
Chemical Name or Material Water
Show More Show Less