Water Hardness Indicator, 0.5% (w/v) Eriochrome Black T in 2-Methoxyethanol, Ricca Chemical

$67.04

Chemical Identifiers

Chemical Name or Material Water Hardness Indicator
Name Note 0.5% (w/v) Eriochrome Black T in 2-Methoxyethanol
CAS [109-86-4, 98.00-100.00%], [1787-61-7, 0.47-0.49%]
Physical Form Dark purple liquid
View More Specs

  • Products 1
Products
Catalog Number Mfr. No. Description Price Quantity Availability  

930032

Ricca Chemical
9300-32
Glass amber; 1L
Each for $67.04
Check Availability
Add to Cart
Descriptions & Specifications
Safety and Handling

Related Products