Alfa Aesar™ Water, LC-MS Grade

Mobile phase solvent in LC-MS analysis

Manufacturer: Alfa Aesar™ 47146K7

Quantity 4L

 View more versions of this product

Catalog No. AA47146K7

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7732-18-5
    Formula Weight 18.02
    MDL Number MFCD00011332
    Beilstein 2050024
    Merck Index 14,10039
    Chemical Name or Material Water
    Grade LC/MS Grade

    Specifications

    Formula Weight 18.02
    TSCA TSCA
    MDL Number MFCD00011332
    CAS 7732-18-5
    Merck Index 14,10039
    Melting Point 0°C
    Density 1.000g/mL
    Color Colorless
    Boiling Point 100°C
    Quantity 4L
    Refractive Index 1.34
    Physical Form Liquid
    Beilstein 2050024
    Odor Odorless
    RTECS Number ZC0110000
    Chemical Name or Material Water
    Grade LC/MS Grade

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit