Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, LCMS, Spectrum™ Chemical
SDP

Click to view available options
Quantity:
4 L
Packaging:
Poly Bottle
CAS 7732-18-5
CAS Max % 1
Packaging Poly Bottle
Residue after Evaporation 1 mg/L
Quantity 4 L
Refractive Index 1.3325-1.3335
Grade High Purity Solvent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.