Learn More
LaMotte Water Quality Classroom Studies
Reinforce key concepts on water treatment and environmental studies.
$57.00
Specifications
For Use With (Application) | Water quality studies |
---|---|
Sufficient For | 25 demonstrations |
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
S66983
![]() |
LaMotte
5963 |
Dissolved Oxygen Lab |
Each for $57.00
|
|
S66988
![]() |
LaMotte
5967 |
Tapwater Hardness Lab |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
View substitute products » |
S66991
![]() |
LaMotte
5962 |
Water Treatment Lab |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
View substitute products » |
Description
- Teacher's manual has tips for organizing the demonstration, test factor information and suggestions for additional experiments
- Also include introductory information, reproducible student procedures/data sheets, lesson plan and reagent composition information
Dissolved Oxygen Lab: Discover the relationship between temperature and oxygen saturation.
Nutrients in Water Lab: Nitrate and phosphate have a key role in indicating pollution. Prepared samples include levels of these nutrients for measuring and discussion.
Tapwater Hardness Lab: Provided sample demonstrates the quantitative and aesthetic qualities of drinking water.
Water Treatment Lab: Investigates the water treatment process by examining the odor, color and turbidity of drinking water.
Order Info
Grades 5 to 8. Class size: 25 demonstrations.
Specifications
Water quality studies | |
25 demonstrations |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.