Learn More
LaMotte Water Quality Educator and Monitoring Outfit
Ideal for long-term water-quality monitoring.
$147.00 - $437.00
Specifications
For Use With (Application) | Long term water quality monitoring |
---|
Includes:
- CD-ROM benchmarks and compares data for each test factor, explains typical causes and effects of higher and lower quality levels
- The Monitor's Handbook (71 pages)
- 100 tests for pH
- 50 tests for the following five factors: nitrate-nitrogen, phosphate, dissolved oxygen, total alkalinity, turbidity, unlimited temperature tests, aquatic macroinvertebrate bioassessment
- Lesson plans for grades 5 to 8 and 9 to 12
Catalog Number | Mfr. No. | Product Type | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Includes | Price | Quantity & Availability | ||||
S66223
![]() |
LaMotte
587001 |
Water Quality Educator and Monitoring Outfit | pH, nitrate-nitrogen, phosphate, dissolved oxygen, alkalinity, total, turbidity, temperature test factor, water quality educator CD ROM and the monitor's handbook, 100 tests for pH, unlimited temperature tests, aquatic macro invertebrate bioassessment |
Each for $437.00
|
|
S66224
![]() |
LaMotte
R5870 |
Reagent Refill | Set of refill reagents as originally supplied in the complete kit |
Each for $147.00
|
|
S66225
![]() |
LaMotte
5872 |
Water Quality Educator CD | Water quality educator CD-ROM, the monitor's handbook, aquatic macro invertebrate bioassessment |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
- Tests for seven basic water-quality test factors Prepares students for field tests or classroom work
- Set up a water-quality monitoring program, procure measurements, and interpret results
- Soft-sided tote made of durable fabric, solid rubber bottom ready for field work; handy side pockets hold thermometer, pens, writing tablets and other tools
Order Info
System requirements:
- Windows: 64MB available RAM, Windows 98, 2000, XP or Vista
- Macintosh: 128MB available RAM, OSX 10.1.5, 10.2.6 or 10.3
Specifications
Long term water quality monitoring |