Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cellucap Water Repellent SMS Coats
Click to view available options
Construction:
1.25 oz. Spunbond Polypropylene
2-ply Spunbond Polypropylene, inner layer of Meltblown
Polycoated Polypropylene
Spunbond Polypropylene, Meltblown
Size:
Large
X-Large
2X-Large
3X-Large
Specifications
Construction | 1.25 oz. Spunbond Polypropylene |
Size | 2X-Large |
Material | Polypropylene |
Color | White |
Closure Type | Snaps |
Cuff Style | Open |
For Use With (Application) | For dusty areas and superb for splash protection |