Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Water Resistant Cryogenic Gloves - Shoulder Length
SDP

Catalog Number p-7163243
Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

National Safety Apparel™ Water Resistant Cryogenic Gloves provide protection from ultra-cold environments in research labs, frozen food processing, dry ice blasting, and more.

  • Gloves offer protection from ultra-cold environments in a wide variety of applications
  • Teflon™ Laminated Nylon with 3M™ Scotchlite™ Thinsulate™ liner
  • Water resistant, breathable membrane for additional splash protection
  • Made in the USA
Description Shoulder Length
Color Blue
Hand Specific
Material Nylon
Product Type Cryogenic Gloves
Size Small
For Use With (Application) Ultra-Cold Environments such as Research Labs, Frozen Food Processing, and Dry Ice Blasting
Liner Material 3M™ Thinsulate™
Series G99CRBER_ _SH
Length (English) 26 in.
Length (Metric) 66 cm
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit