Learn More
Water, RNase, DNase-free, Thermo Scientific Chemicals
Description
Water, RNase, DNase-free high quality, RNase/DNase-free water is used in all molecular biology applications like PCR, DNA. Diethylpyrocarbonate (DEPC)-treated Water is guaranteed nuclease-free (tested for both DNase and RNase) and is suitable for all RNA work.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Water, RNase, DNase-free high quality, RNase/DNase-free water is used in all molecular biology applications like PCR, DNA. Diethylpyrocarbonate (DEPC)-treated Water is guaranteed nuclease-free (tested for both DNase and RNase) and i
Specifications
Color | Colorless |
Boiling Point | 100°C |
Beilstein | 2050024 |
Merck Index | 14,10039 |
Solubility Information | It is soluble in water. |
Formula Weight | 18.02 |
Odor | Odorless |
Density | 1.000 g/mL |
Quantity | 3.8 L |
Physical Form | Liquid |
Show More |
Safety and Handling
EINECSNumber | 231-791-2 |
RTECSNumber | ZC0110000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.