Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, for spectroscopy ACS, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
Packaging:
Glass bottle
7732-18-5
H2O
dihydrogen oxide,dihydrogen monoxide
7732-18-5
H2O
dihydrogen oxide,dihydrogen monoxide
For UV/VIS spectroscopy
Absorbance 0.005 max. at 254nm,0.005 max. at 300nm,0.01 max. at 200nm,0.01 max. at 210nm,Passes Test
Chloride 0.4ppm max.
Linear Formula H2O
Density 1.0000g/mL
Formula Weight 18.02
Physical Form Liquid
Grade ACS
Gradient Analysis Passes Test
Heavy Metals (as Pb) 0.01ppm max.
Nitrate and Nitrite 0.4ppm max.
Packaging Glass bottle
Phosphate 1.0ppm max.
Reducing Substances Passes Test
Refractive Index 1.3334
Silicate 0.01ppm max.
Specific Gravity 1
Sulfate 1.0ppm max.
Viscosity 0.89 mPa.s (25°C)
Boiling Point 100.0°C
Color Colorless
Quantity 250mL
Chemical Name or Material Water, For Spectroscopy
Show More Show Less