Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water Standard in Methanol (1mL = 1mg Water/Certified), Fisher Chemical

Click to view available options
Quantity:
1L
Packaging:
Glass Bottle
7732-18-5
H2O
MFCD00011332
Water Standard in Methanol
7732-18-5
H2O
MFCD00011332
Water Standard in Methanol

For standardizing Karl Fischer Reagent

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

Additional Information Vapor Pressure: 128hPa at 20°C
Melting Point -98°C
Boiling Point 64.7°C
Quantity 1L
Color Colorless
Density 1.11 (Air = 1.0)
Equivalence 1mL = 1mg water
Identification Pass Test
Packaging Glass Bottle
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Molecular Weight (g/mol) 18.02
PubChem CID 962
ChEBI CHEBI:15377
Physical Form Liquid
Grade Certified
Show More Show Less

DANGER!

Emergency Overview
Flammable liquid and vapor. Cannot be made non-poisonous. Vapor harmful. Toxic by inhalation, in contact with skin and if swallowed. Irritating to eyes and skin. May cause irritation of respiratory tract. Toxic: danger of very serious irreversible effects through inhalation, in contact with skin and if s. Explosive. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required.

NFPA

Health:1
Flammability:3
Instability:0