Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, Sterile, Nuclease Free, Low Endotoxin, OmniPur™, Calbiochem™,

Click to view available options
Quantity:
100mL
Packaging:
Plastic Bottle
7732-18-5
H2O
18.015
XLYOFNOQVPJJNP-UHFFFAOYSA-N
dihydrogen oxide,dihydrogen monoxide
962
CHEBI:15377
oxidane
O
7732-18-5
H2O
18.015
XLYOFNOQVPJJNP-UHFFFAOYSA-N
dihydrogen oxide,dihydrogen monoxide
962
CHEBI:15377
oxidane
O

Recommended Storage

Ok to freeze

Physical Form Liquid
Grade Molecular Biology
Packaging Plastic Bottle
Color Colorless
Quantity 100mL