Learn More
Water Turbine
Explore hydroelectric power in the classroom using man's first way of generating electricity.
Supplier: Science First 6112235
Includes: Turbine, all necessary tubing, small color wheel (turns white when turbine reaches optimal speed), instructions
Description
A small wheel is easily spun using water from a faucet.
- Falling water turns potential into kinetic energy
- Kinetic energy in turn drives a generator and produces up to 6VDC
- Turbine constructed of plastic and brass
- Resistant to water damage
Specifications
Water Turbine | |
Turbine, all necessary tubing, small color wheel (turns white when turbine reaches optimal speed), instructions | |
Explores hydroelectric power in the classroom using man's first way of generating electricity | |
Physics |