Water, ultrapure, Spectrophotometric Grade, Alfa Aesar™

Maximum level of impurities: Evaporation residue 2ppm, Specific conductance at 25° 1.5µ mhos/cm, SiO2 0.01ppm, Heavy metals (as Pb) 0.01ppm, Substances reducing permanganate P.T.

Manufacturer: Alfa Aesar 19391M1

Quantity 4 x 1L

 View more versions of this product

Catalog No. AA19391M1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7732-18-5
    Physical Form Liquid
    Molecular Formula H2O
    Formula Weight 18.02
    MDL Number MFCD00011332
    Chemical Name or Material Water
    Grade Spectrophotometric grade

    Specifications

    Color Clear colorless
    Physical Form Liquid
    Formula Weight 18.02
    Quantity 4 x 1L
    MDL Number MFCD00011332
    Molecular Formula H2O
    CAS 7732-18-5
    Appearance Colorless liquid
    Vapor Pressure 23 hPa (17mm Hg) at 20°C
    Chemical Name or Material Water
    Grade Spectrophotometric grade

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit