Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Water, WFI Quality, Sterile Purified Water, Cell Culture Tested, OmniPur™, Calbiochem™,

Click to view available options
Quantity:
500mL
1L
5L
10L
20L
200L
7732-18-5
H2O
18.015
XLYOFNOQVPJJNP-UHFFFAOYSA-N
dihydrogen oxide,dihydrogen monoxide
962
CHEBI:15377
oxidane
O
7732-18-5
H2O
18.015
XLYOFNOQVPJJNP-UHFFFAOYSA-N
dihydrogen oxide,dihydrogen monoxide
962
CHEBI:15377
oxidane
O
Physical Form Liquid
Color Colorless
Quantity 5L