Water, World/GMP, USP/EP/JP Grade, PHARMCO-AAPER

Manufacturer: Pharmco Products 232WORLDDM55UV

Packaging UV poly drum
Quantity 55 gal.

 View more versions of this product

Catalog No. 16-101-403

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7732-18-5
    Physical Form Liquid
    Molecular Formula H2O
    Formula Weight 18.02
    Chemical Name or Material Water

    Specifications

    Formula Weight 18.02
    Quantity 55 gal.
    Color Colorless
    Physical Form Liquid
    Description General purpose solvent use
    Molecular Formula H2O
    CAS 7732-18-5
    Packaging UV poly drum
    Stability Stable under recommended storage conditions.
    Chemical Name or Material Water

    Order Info

    Minimum Order Quantity: 5

    Minimum Order Quantity: 5

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit