Learn More
M+A Matting WaterHog™ Classic Tile Mat
WaterHog™ mats are durable, stain-resistant mats with a unique bi-level design that traps dirt and moisture, keeping floors clean, dry, and safe.
$300.50
Specifications
Certifications/Compliance NFSI Certified. Made in USA
Quantity 1 Ea.
Thickness (English) 7/16 in.
Color Medium Gray
Length (English) 18 in.
View More Specs
Products 2
Catalog Number Mfr. No. Style Price Quantity & Availability  
Catalog Number Mfr. No. Style Price Quantity & Availability  
17-555-123
View Documents
M+A Matting
210570716
Square pattern
Each for $300.50
Add to cart
17-555-124
View Documents
M+A Matting
219570716
Diamond pattern
Each for $300.50
Add to cart
Description
Description
  • Bi-level surface effectively scrapes dirt and moisture from shoes, trapping it beneath shoe level to prevent tracking
  • Reinforced rubber nubs prevent pile from crushing and extend the performance life of the mat
  • Solution-dyed polypropylene fabric is resistant to staining, dries quickly, and will not fade or rot
  • Certified high-traction by the National Floor Safety Institute (NFSI)
  • Available in 18 colors with choice of 4 patterns: waffle, diamond, diagonal, or geometric
  • Available with choice of 100-mil or 200-mil SBR rubber
Specifications
Specifications
NFSI Certified. Made in USA
7/16 in.
18 in.
1 Ea.
Medium Gray