Learn More
M+A Matting WaterHog™ Squares Fashion Mat, Medium Gray
WaterHog™ mats are durable and stain-resistant with a unique bi-level design that traps dirt and moisture, keeping floors clean, dry, and safe.
$37.00 - $901.00
Specifications
Certifications/Compliance | NFSI Certified. Made in USA |
---|---|
Color | Medium Gray |
Quantity | 1 Ea. |
Thickness (English) | 3/8 in. |
Catalog Number | Mfr. No. | Length (English) | Width (English) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (English) | Width (English) | Price | Quantity & Availability | ||||
17-555-661
|
M+A Matting
280571827070 |
18 in. | 27 in. |
Each for $37.00
|
|
||||
17-555-649
|
M+A Matting
2805700203100 |
2 ft. | 3 ft. |
Each for $65.80
|
|
||||
17-555-650
|
M+A Matting
280570034070 |
3 ft. | 4 ft. |
Each for $130.60
|
|
||||
17-555-651
|
M+A Matting
280570035070 |
3 ft. | 5 ft. |
Each for $164.30
|
|
||||
17-555-652
|
M+A Matting
280570038070 |
3 ft. | 8 ft. |
Each for $302.00
|
|
||||
17-555-656
|
M+A Matting
280570310070 |
3 ft. | 10 ft. |
Each for $327.50
|
|
||||
17-555-657
|
M+A Matting
280570312070 |
3 ft. | 12 ft. |
Each for $453.00
|
|
||||
17-555-653
|
M+A Matting
280570046070 |
4 ft. | 6 ft. |
Each for $260.50
|
|
||||
17-555-654
|
M+A Matting
280570048070 |
4 ft. | 8 ft. |
Each for $402.00
|
|
||||
17-555-658
|
M+A Matting
280570410070 |
4 ft. | 10 ft. |
Each for $436.00
|
|
||||
17-555-659
|
M+A Matting
280570412070 |
4 ft. | 12 ft. |
Each for $601.00
|
|
||||
17-555-655
|
M+A Matting
280570068070 |
6 ft. | 8 ft. |
Each for $601.00
|
|
||||
17-555-660
|
M+A Matting
280570612070 |
6 ft. | 12 ft. |
Each for $901.00
|
|
||||
Description
- Bi-level surface effectively scrapes dirt and moisture from shoes, trapping it beneath shoe level to prevent tracking
- Water dam border holds moisture keeping it off your floors (WaterHog mats can hold up to 1.5 gallons of water per square yard)
- Reinforced rubber nubs prevent pile from crushing and extend the performance life of the mat
- Beveled edges provide a safe transition from floor to mat
- Certified high-traction by the National Floor Safety Institute (NFSI)
- PET fabric is made from 100% post-consumer recycled plastic from drink bottles, and SBR rubber backing contains 20% recycled rubber from car tires
- Available in 19 colors
- Feature 78 mil SBR rubber backing
Specifications
NFSI Certified. Made in USA | |
Medium Gray | |
1 Ea. | |
3/8 in. |