Waterjel™ First-aid Sprays

Manufacturer: Moore Medical Corp 63940

 View more versions of this product

Catalog No. 19-898-310

  Check Availability
Add to Cart

    Specifications

    Color Clear
    Composition Benzalkonium Chloride
    For Use With (Application) Prevent infection in minor cuts, scrapes and burns
    Quantity Each
    Volume (English) 2 oz.
    Volume (Metric) 59mL
    Product Type Antiseptic Spray
    pH 6 to 7.5

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit