Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ WaterPro™ BT Water Purification Systems Products for Science Education

The WaterPro BT System is ideal for laboratories needing 1-10L/day, delivering both ultrapure Type I, up to 18.2 megohm-cm, and reverse osmosis (RO)-purified Type III water. Benchtop design is compact, lightweight and quick to install, WaterPro BT may be easily transported and shared among labs.

Manufacturer:  Labconco™ 9015020/EMD

 View more versions of this product

Catalog No. S28405


Add to cart

Requires : WaterPro BT Filter Pack (9019200)

Description

Description

  • Dispensing valve that dispenses >0.5L/min. Type I water by pressing dispense button
  • RO-purified water storage tank 6L capacity
  • Feedwater inlet tubing, 2.13m (7 ft.) long, with 1.27cm (0.5 in.) O.D. Male NPTM, BSPM or GAZ fitting and pre-filter screen
  • RO reject tubing, 203.2cm (80 in.) long
  • Distribution pump and continuous/intermittent recirculation
  • Visual alarms including filter cartridge change after set time has expired or amount of water has been reached, resistivity less than setpoint, incorrect filter installation, and water detected in tank while in Flush mode
  • Keypad for accessing display settings, function settings, and calibrations
  • Rear-mounted tank outlet valve releases RO water from tank
  • Maximum noise level (50dB) at distance of 1m (3.25 ft.)
  • Thermoplastic housing with removable thermoplastic cover
  • Optional 185/254nm UV lamp to remove bacteria and certain organics
  • Optional remote dispenser to allow WaterPro BT to be placed up to 2m (6.5ft.) away from dispensing location
  • Delivers ultrapure Type I, up to 18.2 megohm-cm at typical rate of >0.5L/min. at inlet water temperature of 25°C
  • Delivers reverse osmosis (RO)-purified Type III water at rate up to 3L/hour (0.05L/min.)
Feedwater requirements - Type I water:
  • Maximum inlet pressure - 87.0psi (6 bar)
  • Minimum inlet pressure - 7.3psi (0.5 bar)
  • Minimum Inlet Flow rate - 40L/hour (10.6 gal./hour)
UV Model:
  • UV Lamp, 185 and 254nm
Remote Dispense Model:
  • Remote Dispenser with 2m (6.5 ft.) tether
  • White powder-coated steel dispenser stand
Specifications

Specifications

21.2 in.
11.4 in.
16.6 in.
UL 61010-1; CAN/CSA C22.2 No. 61010.1; CE electrical safety and electromagnetic compatibility
115V, 60Hz, 2A
115V, 15A
53.8cm
29cm
42.1cm
Type I water: Instrumental trace element analysis, mixing standardized acid/base solutions, and media solutions Type III water: Non-critical laboratory applications such as reagent preparation and glassware rinsing
N/A
Water Purification System
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit