Learn More
Oakton™ Waterproof CON 450 Portable Conductivity Meter
Rugged, reliable and accurate meter at an economical price!
 View more versions of this product
Catalog No. 15-500-129
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
CON 450 Meter with Probe and Calibration
CON 450
0 to 200mS
±1 % full-scale
0.01ppm to 0.1 ppt
0 to 50 ppt
Manual/Automatic
0.1
Backlit LCD
Two AA batteries or optional universal power adapter
Two AA batteries
CE marked, TUV 3-1, FCC Class A
One point per range
Ideal for any environment like manufacturing floor, field testing, and the laboratory.
Three years
Meter, Conductivity cell (K: 1.0), Grip-Clip holder, AA batteries and NIST-Traceable calibration
0.01μS to 0.1mS
0 to 200 ppt
±1% full-scale
±1% full-scale
-10° to +110°C (17° to 230°F)
±0.5°C (0.9°F)
500
500 hours
IP67
19.8 x 8 x 5.7cm (7.8 x 3.12 x 2.25 in.)
0.1/1.0/10.0