Learn More
Thermo Scientific™ Waterproof Cryo Gloves Available on GSA/VA Contract for Federal Government customers only.
Safely handle samples from ultra low and cryogenic storage with Thermo Scientific? Waterproof Cryo Gloves, available in mid-arm, elbow-length and shoulder-length options.
Manufacturer:  Thermo Scientific™ 189446
 View more versions of this product
Catalog No. 19-807-668
  Check Availability
Add to cart
Specifications
Description
Safely handle samples from ultra low and cryogenic storage with Thermo Scientific™ Waterproof Cryo Gloves, available in mid-arm, elbow-length and shoulder-length options.
  • Provides splash protection
  • Comfortable, lightweight, breathable
  • Side release buckles for quick on/off
Description & Specifications
Specifications
Cryogenic Preservation
Waterproof Cryo Gloves
45.72cm
Large
18 in.