Learn More
Oakton™ Waterproof DO 450 Portable Dissolved Oxygen Meter
Optical RDO technology offers fast response times and long-term stability with minimal maintenance
 View more versions of this product
Catalog No. 15-500-132
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
DO 450 Meter w/Optical Probe
DO 450 meter, RDO probe, RDO sensor cap and two AA batteries
±0.5°C (±0.9°F)
500 sets
Two AA batteries
CE marked, TUV 3-1, FCC Class A
±2%
-10° to +110°C
Three years
0.1
Backlit LCD
500 hours
IP67
20 x 8 x 5.7cm (7.9 x 3.12 x 2.25 in.)
17° to 230°F