Oakton™ Waterproof PC 450 Portable Multiparameter Meter

Choose meter with combination pH/conductivity/temperature probe or separate pH and conductivity/temperature probes

 View more versions of this product

Catalog No. 15-500-138

  Check Availability
Add to Cart

    Specifications

    Temperature (Metric) Range -10° to +110°C
    Product Type Multiparameter Meter with separate pH and conductivity/ATC probes
    Temperature (English) Range 17° to 230°F
    Width (English) 3.12 in.
    Accuracy (Conductivity) ±1% full-scale
    Range (TDS) 0 to 200 ppt
    Resolution (Temperature) ±0.5°C (±0.9°F)
    Data Points 150 sets
    For Use With (Application) Ideal for manufacturing floor, field testing, as well as the laboratory.
    Resolution (mV) 0.1 to 1
    Accuracy (mV) ±0.5
    Resolution (Salinity) 0.1 ppt
    Accuracy (pH) ±0.01
    Length (English) 7.9 in.
    Length (Metric) 20cm
    Width (Metric) 8cm
    Range (Conductivity) 0 to 200mS
    Resolution (Conductivity) 0.01μS to 0.1mS
    Resolution (TDS) 0.01ppm to 0.1 ppt
    Accuracy (TDS) ±1% full-scale
    Range (Salinity) 0 to 50 ppt
    Accuracy (Salinity) ±1% full-scale
    Includes Meter, pH probe, conductivity/temperature cell K=1.0, Grip-Clip holder and two AA batteries
    Calibration Points (Conductivity) One calibration point per range
    Range (Temperature) 0.1
    Resolution (pH) 0.01
    Display Type Backlit LCD
    Battery Type Two AA batteries
    IP Rating IP67
    Certifications/Compliance CE marked, TUV 3-1, FCC Class A
    Calibration Points (pH) Up to to six points from USA, NIST, DIN, and custom pH buffer sets
    Height (Metric) 5.7cm
    Range (pH) -2.00 to 16.0
    Range (mV) ±2000
    Height (English) 2.25 in.
    Warranty Three years

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit