Learn More
Oakton™ Waterproof PC 450 Portable Multiparameter Meter
Choose meter with combination pH/conductivity/temperature probe or separate pH and conductivity/temperature probes
 View more versions of this product
Catalog No. 15-500-139
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Meter, pH probe, conductivity/temperature cell K=1.0, Grip-Clip holder, two AA batteries and NIST-Traceable calibration certificate provided by InnoCal
0.01
±2000
±0.5
0.01μS to 0.1mS
0 to 200 ppt
±1% full-scale
0.1 ppt
One calibration point per range
±0.5°C (±0.9°F)
150 sets
IP67
Up to to six points from USA, NIST, DIN, and custom pH buffer sets
2.25 in.
7.9 in.
17° to 230°F
3.12 in.
Three years
-2.00 to 16.0
±0.01
0.1 to 1
0 to 200mS
±1% full-scale
0.01ppm to 0.1 ppt
0 to 50 ppt
±1% full-scale
0.1
Backlit LCD
Two AA batteries
CE marked, TUV 3-1, FCC Class A
Ideal for manufacturing floor, field testing, as well as the laboratory.
5.7cm
20cm
-10° to +110°C
8cm
Multiparameter Meter with separate pH and conductivity/ATC probes and Calibration