Oakton™ Waterproof PC 450 Portable Multiparameter Meter Kit

Choose meter with combination pH/conductivity/temperature probe or separate pH and conductivity/temperature probes

 View more versions of this product

Catalog No. 15-500-140

  Check Availability
Add to Cart

    Specifications

    Temperature (English) Range 17° to 230°F
    Width (Metric) 8cm
    Calibration Points (Conductivity) One calibration point per range
    Battery Type Two AA batteries
    IP Rating IP67
    Calibration Points (pH) Up to to six points from USA, NIST, DIN, and custom pH buffer sets
    Length (English) 7.9 in.
    Length (Metric) 20cm
    Product Type Multiparameter Meter Kit with Combination Probes
    Warranty Three years
    Resolution (TDS) 0.01ppm to 0.1 ppt
    Accuracy (TDS) ±1% full-scale
    Range (Salinity) 0 to 50 ppt
    Resolution (Salinity) 0.1 ppt
    Accuracy (Salinity) ±1%full-scale
    Accuracy (Temperature) ±0.5°C (±0.9°F)
    Resolution (pH) 0.01
    Accuracy (pH) ±0.01
    Range (mV) ±2000
    Width (English) 3.12 in.
    Includes Meter, combination pH/conductivity/temperature probe, calibration solution, two AA batteries and hard carrying case
    Range (pH) -2.00 to 16.0
    For Use With (Application) Ideal for manufacturing floor, field testing, as well as the laboratory.
    Height (English) 2.25 in.
    Height (Metric) 5.7cm
    Temperature (Metric) Range -10° to +110°C
    Range (TDS) 0 to 200 ppt
    Resolution (Temperature) 0.1
    Display Type Backlit LCD
    Data Points 150 sets
    Certifications/Compliance CE marked, TUV 3-1, FCC Class A
    Resolution (mV) 0.1 to 1
    Accuracy (mV) ±0.5
    Range (Conductivity) 0 to 200mS
    Resolution (Conductivity) 0.01μS to 0.1mS
    Accuracy (Conductivity) ±1% full-scale

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit