Learn More
Oakton™ Waterproof PD 450 Portable pH/DO Meter
One meter to measure both pH and Dissolved Oxygen of samples
 View more versions of this product
Catalog No. 15-500-145
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
PD 450 pH/DO Meter with Probes and Calibration
PD 450 meter, pH probe, RDO probe, sensor cap, two AA batteries and NIST-traceable calibration certificate provided by InnoCal
0.01
±2000
±0.5% full scale
0.1 ppm; 0.1%
Up to six points
±0.5
500 sets
IP67
Ideal for plant or field testing.
6.3cm
20cm
-10° to +110°C
8cm
Three years
-2.00 to 16.00
±0.01
0.01
0 to 20ppm; 0 to 200%
±2%
0.1
Backlit LCD
Two AA batteries
CE marked, TUV 3-1, FCC Class A
2.5 in.
7.9 in.
17° to 230°F
3.12 in.