Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Waterproof Cryogenic Gloves - Shoulder Length
SDP

National Safety Apparel™ Waterproof Cryogenic Gloves feature a breathable membrane for additional splash protection.

$561.00

Specifications

Color Blue
Description Shoulder Length
For Use With (Application) Cryogenic Liquids/Gases
Length (English) 26 in.
Liner Material 3M™ Thinsulate™
View More Specs

Products 3
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
17-355-068
SDP
View Documents
National Safety Apparel™
G99CRBEPMDSH
Medium
Pair for $561.00
Add to cart
 
17-355-069
SDP
View Documents
National Safety Apparel™
G99CRBEPLGSH
Large
Pair for $561.00
Add to cart
 
17-355-070
SDP
View Documents
National Safety Apparel™
G99CRBEPXLSH
X-Large
Pair for $561.00
Add to cart
 
Description

Description

  • Teflon™ Laminated Nylon with 3M™ Scotchlite™ Thinsulate™ liner
  • Waterproof and breathable membrane for additional splash protection
  • Made in the USA
Specifications

Specifications

Blue
Cryogenic Liquids/Gases
3M™ Thinsulate™
Waterproof Cryogenic Gloves
Shoulder Length
26 in.
Nylon
G99CRBEP_ _SH
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit