Learn More
Waters Corp Spherisorb C8 Column, 80Å, 3 µm, 4.6 mm X 100 mm, 1/pk
Supplier:  Waters Corp PSS832212
Encompass_Preferred
Spherisorb C8 columns are general purpose, silica-based reversed-phase columns that contain an Octyl ligand. These USP L7 columns are used when you require a less retentive sorbent compared to the Spheriorb ODS columns.
Catalog No. 50-784-413