Learn More
LaMotte Watershed Tour
Kit guides the class through a week-long series of activities focused on stream and river ecosystems.
$11.20 - $104.00
Specifications
Min. Grade Level | 4 |
---|---|
Max. Grade Level | 8 |
Includes: Lecture materials, illustrated handouts, teacher tips, test procedures, TesTabs reagents, data sheets and games
Description
Designed for teachers who are unable to visit a watershed with their students
- Teaches students about stream ecology, water quality issues and their own connection to a watershed
- Enables students to perform simulated tests on four sites along a river continuum from the headwaters to the mouth of the river as it enters the estuary to study how the river changes and how human activities can influence water quality
Order Info
Grades 4 to 8. Class size: 30 students working in groups.
Specifications
4 | |
8 |