Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Watson-Marlow Manifold Tubing - Marprene

Double pump segment manifold tubing — use one, transfer to the other for double life

Manufacturer:  Watson-Marlow 979.0050.000

 View more versions of this product

Catalog No. 14-283-170


Add to cart

Description

Description

  • Made from autoclavable marprene
  • For use with 400DM2 and 400DM3 pumpheads
  • Can be used with any multichannel cartridges
  • Adjacent cartridges may contain tubing of a different material or size

Specifications

Specifications

Orange/yellow
0.019 in.
Thermoplastic Elastomer
0.8mm
For 401U/DM2 and 401U/DM3
0.013 to 0.92 (four roller pump), 0.012 to 0.830 (eight roller pump)mL/min.
Yes
0.5mm
0.03 in.
Double Segment Manifold Tubing
1Each
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit