Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Watson-Marlow PVC Manifold Tubing

$86.00

Specifications

Color Code Purple
Autoclavable Autoclavable
Material PVC
For Use With Multichannel cassette pumphead
View More Specs

Products 2
Catalog Number Mfr. No. Diameter (English) Inner Diameter (Metric) Inner Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (English) Inner Diameter (Metric) Inner Price Quantity & Availability  
14-284-144
View Documents
Watson-Marlow
980.0238.000
0.09 in. 2.29 mm
Pack of 12 for $86.00
Add to cart
 
14-284-143
View Documents
Watson-Marlow
980.0205.000
0.08 in. 2.05 mm
Pack of 6 for $86.00
Add to cart
 
Description

Description

Specifications

Specifications

Purple
Autoclavable
PVC
Multichannel cassette pumphead
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit