Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Watson-Marlow Single-Segment Manifold Tubing for Peristaltic Pump Models 205S/CA and 205U/CA: Marprene

For use with Peristaltic Pump Models 205S/CA and 205U/CA.

Manufacturer:  Watson-Marlow 978.0025.000

 View more versions of this product

Catalog No. 14-283-74


Add to cart

Description

Description

For use with Peristaltic Pump Models 205S/CA and 205U/CA.

Specifications

Specifications

Orange/Blue
0.25mm
0.03 in.
Single Segment Manifold Tubing
176°F
41°F
0.25mm
0.009 in.
Marprene
0.8mm
205S/CA and 205U/CA peristaltic pump
80°C
5°C
0.001 to 0.265mL/min.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit