Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Single-Segment Manifold Tubing for Peristaltic Pump Models 205S/CA and 205U/CA: Marprene

Manufacturer:  Watson-Marlow Bredel 978.0129.000

 View more versions of this product

Catalog No. 14-283-82


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Marprene
1.29mm
0.03 in.
Grey/Grey
5 to 80°C
1.29mm
Single Segment Manifold Tubing
0.029 to 5.27mL/min.
0.8mm
41 to 176°F
205S/CA and 205U/CA peristaltic pump
0.05 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit