Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Watson-Marlow Single-Segment Manifold Tubing for Peristaltic Pump Models 205S/CA and 205U/CA: Marprene
For use with Peristaltic Pump Models 205S/CA and 205U/CA.
Supplier: Watson-Marlow 978.0205.000
Description
For use with Peristaltic Pump Models 205S/CA and 205U/CA.
Order Info
Comes in packs of six single segments.
Specifications
Single Segment Manifold Tubing | |
2.05 mm | |
0.03 in. | |
205S/CA and 205U/CA peristaltic pump | |
5°C | |
0.69 to 12.4mL/min. | |
176°F |
8/100 in. | |
Marprene | |
0.8 mm | |
41°F | |
2.05mm | |
Purple/Purple | |
80°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction