Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Watson-Marlow Tubing for Peristaltic Pump Models 205S/CA and 205U/CA; Material Viton™ Acid Resistant

Manufacturer:  Watson-Marlow 986.0185.000

 View more versions of this product

Catalog No. 14-283-154


Add to cart

Specifications

Specifications

Green/Green
1.85mm
Manifold Tube Element
0.057 to 10.3mL/min.
7/100 in.
Viton™
205S/CA and 205U/CA peristaltic pump
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit