Learn More
Wave Generators Products for Science Education
Variable or adjustable phase wave generators available to show different wave effects.
$94.00
Specifications
For Use With (Application) To show different wave effects
View More Specs
Products 2
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
S52082 Products for Science Education
View Documents
Science FirstSupplier Diversity Partner
15445
Adjustable Phase Circular and Straight Wave Generator
Each for $94.00
Add to cart
S52121 Products for Science Education
View Documents
Science FirstSupplier Diversity Partner
15490
Variable Phase Circular Wave Generator This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Variable Phase, Two-Point Source
  • Phase relationship can be continuously adjusted from 0° to 360° without interrupting the wave pattern
  • Circular waves from a single point can also be generated
  • Mounted inside or outside the tank
Adjustable Phase, Two-Point Source
  • Phase relationship can be adjusted from 0° to 360°
  • Produce both circular and straight waves of various frequencies
  • Mounted inside or outside the ripple tank
Specifications
Specifications
To show different wave effects