Learn More
ADS Biotec IncSupplier Diversity Partner WAVE Optimized Buffer B, 4 x 2.5 liter bottles DFS Item
Manufacturer:  ADS Biotec IncSupplier Diversity Partner 553402
WAVE Optimized Buffers were specifically developed to provide highly reproducible retention times with WAVE System instrumentation. WAVE Optimized Buffers have a longer shelf life than buffers prepared from concentrate. They will allow you to generate reproducible data from day to day, week to week, system to system and lab to lab. Not only will they save you time, but reduce problems with water quality and maintenance of purification systems.
Catalog No. NC1631328