Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wax Paraffin, White, Cake, Spectrum™

Click to view available options
Quantity:
1 lbs.
12 x 1 lbs.
22.7 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
8002-74-2
C21H27NO3
341.45
JWHAUXFOSRPERK-UHFFFAOYNA-N
1-{2-[2-hydroxy-3-(propylamino)propoxy]phenyl}-3-phenylpropan-1-one
CCCNCC(O)COC1=CC=CC=C1C(=O)CCC1=CC=CC=C1
8002-74-2
C21H27NO3
341.45
JWHAUXFOSRPERK-UHFFFAOYNA-N
1-{2-[2-hydroxy-3-(propylamino)propoxy]phenyl}-3-phenylpropan-1-one
CCCNCC(O)COC1=CC=CC=C1C(=O)CCC1=CC=CC=C1

W1025, 8002-74-2

Spectrum™ Wax Paraffin, White, Cake consists of a mix of hydrocarbon molecules and is derivable from coal, petroleum, or oil shale. It is used in electrical insulation, lubrication and candles. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Packaging Fiber Drum
Quantity 22.7 kg
CAS Max % 1
Grade Reagent