Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WAY 100635 maleate, Tocris Bioscience™

Catalog Number p-4817147
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
634908-75-1
C29H40N4O6
540.661
QYOBEWLYELTWTQ-UHFFFAOYSA-N
N-[2-[4-(2-Methoxyphenyl)-1-piperazinyl]ethyl]-N-2-pyridinylcyclohexanecarboxamide maleate
77518953
formic acid;N-[2-[4-(2-methoxyphenyl)piperazin-1-yl]ethyl]-N-pyridin-2-ylcyclohexanecarboxamide;prop-2-enoic acid
COC1=CC=CC=C1N2CCN(CC2)CCN(C3=CC=CC=N3)C(=O)C4CCCCC4.C=CC(=O)O.C(=O)O
This item is not returnable. View return policy
634908-75-1
C29H40N4O6
540.661
QYOBEWLYELTWTQ-UHFFFAOYSA-N
N-[2-[4-(2-Methoxyphenyl)-1-piperazinyl]ethyl]-N-2-pyridinylcyclohexanecarboxamide maleate
77518953
formic acid;N-[2-[4-(2-methoxyphenyl)piperazin-1-yl]ethyl]-N-pyridin-2-ylcyclohexanecarboxamide;prop-2-enoic acid
COC1=CC=CC=C1N2CCN(CC2)CCN(C3=CC=CC=N3)C(=O)C4CCCCC4.C=CC(=O)O.C(=O)O

Potent 5-HT1A antagonist; also D4 agonist

Potent, silent antagonist of 5-HT1A receptors (IC50 = 2.2 nM; Ki = 0.84 nM for rat 5-HT1A receptors). Displays 100-fold selectivity for 5-HT1A over other 5-HT subtypes. Also exhibits agonist activity at dopamine D4 receptors.
Quantity 10 mg
Formula Weight 538.64
Percent Purity >99%
Chemical Name or Material WAY 100635 maleate
Recommended Storage Desiccate at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit